missing translation for 'onlineSavingsMsg'
Learn More

Melford TEMED

Product Code. p-200087228
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 30018830

Brand: Melford T180000.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin.

Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin. Suitable for electrophoresis and tested as a catalyst for polyacrylamide gel electrophoresis.

Specifications

Chemical Name or Material TEMED
CAS 110-18-9
Quantity 100 mL
Molecular Formula C6H16N2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.