missing translation for 'onlineSavingsMsg'
Learn More

Melford TEMED

Product Code. 30018830
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018830 100 mL 100mL
30018829 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018830 Supplier Melford Supplier No. T180000.1
  • ONLINE EXCLUSIVE PRICE
    £95.45
    £79.92 / 100mL
    Save £15.53
    16% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin.

Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin. Suitable for electrophoresis and tested as a catalyst for polyacrylamide gel electrophoresis.

Specifications

Chemical Name or Material TEMED
CAS 110-18-9
Quantity 100 mL
Molecular Formula C6H16N2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.