missing translation for 'onlineSavingsMsg'
Learn More

Temanogrel, MedChemExpress

Product Code. 16496747
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496747 5 mg 5mg
16451059 1 mg 1mg
16444048 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496747 Supplier Medchem Express Supplier No. HY10560

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Temanogrel is a highly selective 5-HT2A receptor antagonist with a Ki of 4.9 nM.

Temanogrel is a highly selective 5-HT2A receptor antagonist with a Ki of 4.9 nM.

Specifications

Chemical Name or Material Temanogrel
CAS 887936-68-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H28N4O4
Quantity 5 mg
Synonym APD791
Solubility Information DMSO : 125 mg/mL (286.37 mM; Need ultrasonic)
SMILES CN1C(C2=C(OCCN3CCOCC3)C=CC(NC(C4=CC=CC(OC)=C4)=O)=C2)=CC=N1
Molecular Weight (g/mol) 436.5
Formula Weight 436.5
Percent Purity 98.14%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.