missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEM7 Recombinant Rabbit Monoclonal Antibody (104), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£218.00 - £436.00
Specifications
| Antigen | TEM7 |
|---|---|
| Clone | 104 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17769773
|
Invitrogen™
MA543428 |
50 μL |
£218.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17749683
![]() |
Invitrogen™
MA543134 |
100 μL |
£436.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specificity: Human TEM7/PLXDC1
Recently, using SAGE (Serial Analysis of Gene Expression) technology, St. Croix et al, have identified 46 genes, whose expression is specifically elevated in tumor-associated endothelium. Nine of these genes were prominently expressed only in tumor endothelial cells (EC), but were absent or barely detectable in normal ECs, and named as Tumor Endothelial Markers (TEMs, TEM 1-9). TEM7 (Tumor endothelial marker 7) transcripts are specifically expressed in the endothelium of colorectal cancer, primary cancers of lung, pancreas, breast, and brain. TEM7 is expressed specifically in endothelium of these cancers, whether primary or metastasis. The other six members of this family (TEM1, 3, 4, 5, 8, and 9) also show similar expression pattern in lung and brain tumors, and liver metastasis. Since most of the genes expressed differentially in tumor endothelium are also expressed during angiogenesis, these newly discovered genes might provide important resources for basic and clinical studies of human angiogenesis.Specifications
| TEM7 | |
| 1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| 2410003I07Rik; ADP-ribosylation factor-like 12; AI848450; Arl12; DKFZp686F0937; FLJ36270; FLJ45632; plexin domain containing 1; plexin domain-containing protein 1; PLXDC1; TEM3; TEM7; TEM7 related; tumor endothelial marker 3; Tumor endothelial marker 7; tumor endothelial marker7 | |
| PLXDC1 | |
| IgG | |
| Protein A | |
| Antibody |
| 104 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8IUK5 | |
| 57125 | |
| Recombinant Human TEM7/PLXDC1 protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| PLXDC1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title