missing translation for 'onlineSavingsMsg'
Learn More

TEM7/PLXDC1 Antibody (104), DyLight 680 , Novus Biologicals™

Product Code. 18405829 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405829 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405829 Supplier Novus Biologicals Supplier No. NBP289972FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TEM7/PLXDC1 Monoclonal antibody specifically detects TEM7/PLXDC1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEM7/PLXDC1
Applications ELISA
Classification Monoclonal
Clone 104
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human TEM7/PLXDC1 (AAH36059.1; Met 1-Thr 426).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 57125
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.