missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEM5/GPR124 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-77315CL1
This item is not returnable.
View return policy
Description
TEM5/GPR124 Polyclonal antibody specifically detects TEM5/GPR124 in Human,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| TEM5/GPR124 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| DKFZp434C211, DKFZp434J0911, G protein-coupled receptor 124, G-protein coupled receptor 124, KIAA1531FLJ14390, TEM5Tumor endothelial marker 5 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of the human TEM5. The immunogen is located within amino acids 1270 - 1320 of TEM5. Amino Acid Squence: CGALEKESHRRSYPLN | |
| 0.1 mL | |
| Cancer, Endothelial Cell Markers, GPCR | |
| 25960 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction