missing translation for 'onlineSavingsMsg'
Learn More

TEM5/GPR124 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18416573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416573 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416573 Supplier Novus Biologicals Supplier No. NBP177315AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TEM5/GPR124 Polyclonal antibody specifically detects TEM5/GPR124 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEM5/GPR124
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias DKFZp434C211, DKFZp434J0911, G protein-coupled receptor 124, G-protein coupled receptor 124, KIAA1531FLJ14390, TEM5Tumor endothelial marker 5
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of the human TEM5. The immunogen is located within amino acids 1270 - 1320 of TEM5. Amino Acid Squence: CGALEKESHRRSYPLN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endothelial Cell Markers, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 25960
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.