missing translation for 'onlineSavingsMsg'
Learn More

TEM4 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18421454 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421454 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421454 Supplier Novus Biologicals Supplier No. NBP177314IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TEM4 Polyclonal antibody specifically detects TEM4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEM4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias 164 kDa Rho-specific guanine-nucleotide exchange factor, FLJ90019, KIAA0337P164RHOGEF, p164RhoGEF, p164-RhoGEFRho-specific guanine-nucleotide exchange factor 164 kDa, Rho guanine nucleotide exchange factor (GEF) 17, TEM4rho guanine nucleotide exchange factor 17, Tumor endothelial marker 4
Host Species Rabbit
Immunogen Antibody was raised against a 18 amino acid synthetic peptide near the center of human TEM4. The immunogen is located within amino acids 910 - 960 of TEM4. Amino Acid Squence: KKRPARSSHQELRRDEG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9828
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.