missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18160-1-AP-20UL
This item is not returnable.
View return policy
Description
CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.Specifications
| TEM1 | |
| Polyclonal | |
| Unconjugated | |
| CD248 | |
| CD164L1, CD248, CD248 molecule, endosialin, Endosialin, TEM1, Tumor endothelial marker 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 293669, 57124, 70445 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91V98, Q9HCU0 | |
| Cd248 | |
| TEM1 Fusion Protein Ag12990 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction