missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEM1 Mouse anti-Human, Clone: 1F9B4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TEM1 |
|---|---|
| Clone | 1F9B4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16827145
|
Proteintech
60170-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16817145
|
Proteintech
60170-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.Specifications
| TEM1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CD164L1, CD248, CD248 molecule, endosialin, Endosialin, TEM1, Tumor endothelial marker 1 | |
| CD248 | |
| IgG1 | |
| Caprylic Acid Precipitation | |
| Antibody |
| 1F9B4 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9HCU0 | |
| 57124 | |
| TEM1 Fusion Protein Ag13334 | |
| Primary | |
| -20°C | |
| CD248 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title