missing translation for 'onlineSavingsMsg'
Learn More

Telotristat, MedChemExpress

Product Code. 16488227
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488227 5 mg 5mg
16445518 10 mg 10mg
16410196 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488227 Supplier Medchem Express Supplier No. HY13055B
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Telotristat (LP-778902) is a potent tryptophan hydroxylase inhibitor with an in vivo IC50 of 0.028 μM.

Telotristat (LP-778902) is a potent tryptophan hydroxylase inhibitor with an in vivo IC50 of 0.028 μM.

Specifications

Chemical Name or Material Telotristat
CAS 1033805-28-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H22ClF3N6O3
Quantity 5 mg
Synonym LP-778902
Solubility Information DMSO : 33.33 mg/mL (60.94 mM; Need ultrasonic)
SMILES N[C@@H](CC1=CC=C(C2=NC(N)=NC(O[C@H](C3=CC=C(Cl)C=C3N4N=C(C)C=C4)C(F)(F)F)=C2)C=C1)C(O)=O
Molecular Weight (g/mol) 546.93
Formula Weight 546.93
Percent Purity 98.91%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.