missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TELO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116718
This item is not returnable.
View return policy
Description
Positive Samples: K-562, HeLa Immunogen sequence: ASEAGTSLVP ATAEPPAETP AEIVDGGVPQ AQLAGSDSDL DSDDEFVPYD MSGDRELKSS KAPAYVRDCV EALTTSEDIE RWEAALRALE GLVYRSPTAT R.
TELO2 is a protein that functions as an S-phase checkpoint protein in the cell cycle. The protein may also play a role in DNA repair.
Specifications
| TELO2 | |
| Polyclonal | |
| Unconjugated | |
| TELO2 | |
| 1200003M09Rik; AI415602; CLK2; hCLK2; KIAA0683; mKIAA0683; protein clk-2 homolog; RGD1305553; Tel2; TEL2, telomere maintenance 2, homolog; TEL2, telomere maintenance 2, homolog (S. cerevisiae); Telo2; Telomere length regulation protein TEL2 homolog; telomere maintenance 2; YHFS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 71718, 9894 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.69 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9DC40, Q9Y4R8 | |
| TELO2 | |
| A synthetic peptide corresponding to a sequence within amino acids 450-550 of human TELO2 (NP_0571952). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction