missing translation for 'onlineSavingsMsg'
Learn More

Telmisartan tert-Butyl Ester, TRC

Product Code. 30011592
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011592 1 g 1g
30006111 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011592 Supplier Toronto Research Chemicals Supplier No. TRCT0170301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144702-26-1
Molecular Formula C37 H38 N4 O2
Molecular Weight (g/mol) 570.72
Synonym 1,1-Dimethylethyl 4'-[(1,4'-dimethyl-2'-propyl[2,6'-bi-1H-benzimidazol]-1'-yl)methyl][1,1'-biphenyl]-2-carboxylate, [1,1'-Biphenyl]-2-carboxylic acid, 4'-[(1,4'-dimethyl-2'-propyl[2,6'-bi-1H-benzimidazol]-1'-yl)methyl]-, 1,1-dimethylethyl ester, Telmisartan tert-Butyl Ester, Telmisartan Imp C (EP)
IUPAC Name tert-butyl 2-[4-[[4-methyl-6-(1-methylbenzimidazol-2-yl)-2-propylbenzimidazol-1-yl]methyl]phenyl]benzoate
SMILES CCCc1nc2c(C)cc(cc2n1Cc3ccc(cc3)c4ccccc4C(=O)OC(C)(C)C)c5nc6ccccc6n5C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C37H38N4O2/c1-7-12-33-39-34-24(2)21-27(35-38-30-15-10-11-16-31(30)40(35)6)22-32(34)41(33)23-25-17-19-26(20-18-25)28-13-8-9-14-29(28)36(42)43-37(3,4)5/h8-11,13-22H,7,12,23H2,1-6H3
Formula Weight 570.2995
Percent Purity >95%
Chemical Name or Material Telmisartan tert-Butyl Ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.