missing translation for 'onlineSavingsMsg'
Learn More

Telmisartan, MedChemExpress

Product Code. 16492077
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492077 50 mg 50mg
16486366 100 mg 100mg
16411716 500 mg 500mg
16471389 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16492077 Supplier Medchem Express Supplier No. HY13955
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Telmisartan is a potent, long lasting antagonist of angiotensin II type 1 receptor (AT1), selectively inhibiting the binding of 125I-AngII to AT1 receptors with IC50 of 9.2 nM.

Telmisartan is a potent, long lasting antagonist of angiotensin II type 1 receptor (AT1), selectively inhibiting the binding of 125I-AngII to AT1 receptors with IC50 of 9.2 nM.

Specifications

Chemical Name or Material Telmisartan
CAS 144701-48-4
Color White
Purity Grade Notes Research
Molecular Formula C33H30N4O2
Quantity 50 mg
Synonym BIBR 277
Solubility Information DMSO : 6.67 mg/mL (12.96 mM; Need ultrasonic)
SMILES O=C(C1=CC=CC=C1C2=CC=C(CN3C4=CC(C5=NC6=CC=CC=C6N5C)=CC(C)=C4N=C3CCC)C=C2)O
Molecular Weight (g/mol) 514.62
Formula Weight 514.62
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.