missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tellurium solution for ICP/MS 10mg/L HNO3 matrix, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Specifications
Specifications
| Packaging | Bottle |
| CAS | 7697-37-2 |
| Molecular Formula | HNO3 |
| Concentration | 10 mg/L |
| MDL Number | MFCD00011349 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| SMILES | O[N+]([O-])=O |
| IUPAC Name | nitric acid |
| Solvent or Matrix | HNO3 |
| Molecular Weight (g/mol) | 63.01 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction