missing translation for 'onlineSavingsMsg'
Learn More

Tellurium, plasma standard solution, Specpure™ Te 10,000μg/mL

Product Code. 15467455
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
This item is not returnable. View return policy

Product Code. 15467455

Brand: Thermo Scientific Alfa Aesar 047187.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00134062

Specifications

Odor Acidic
Physical Form Liquid
Concentration Te in 30% HCl
UN Number UN1789
Quantity 100 mL
Grade Specpure
Chemical Name or Material Tellurium, plasma standard solution, Te 10,000μg/mL

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.