missing translation for 'onlineSavingsMsg'
Learn More

Tellurium lump, typically 12mm (0.47 in.), 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11396847
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
13494-80-9
Te
127.60
MFCD00134062
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
6327182
CHEBI:30452
tellurium
[Te]
This item is not returnable. View return policy
13494-80-9
Te
127.60
MFCD00134062
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
6327182
CHEBI:30452
tellurium
[Te]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 g
Percent Purity 99.9999%
Odor Odorless
Form Lump
Assay (metals basis)
Chemical Name or Material Tellurium lump, Typically 12mm (0.47 in.)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'