missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) bromide, ultra dry, 99.999% (metals basis)

Product Code. 11365737
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365737 250 g 250g
11375737 10 g 10g
11385737 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11365737 Supplier Thermo Scientific Alfa Aesar Supplier No. 043439.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10031-27-3 | Br4Te | 447.22 g/mol

Tellurium tetrabromide it acts as a chemical and organic intermediate. It is also acts a conductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-27-3
Molecular Formula Br4Te
Molecular Weight (g/mol) 447.22
MDL Number MFCD00049575
InChI Key PTYIPBNVDTYPIO-UHFFFAOYSA-N
Synonym tellurium tetrabromide, tellurium iv bromide, tellurium bromide tebr4 , t-4, unii-7a29efj1af, 7a29efj1af, tetrabromo-, e4-tellane, tebr4, acmc-1bozh, tellurium iv bromide, ultra dry
PubChem CID 82311
IUPAC Name tetrabromo-λ⁴-tellane
SMILES Br[Te](Br)(Br)Br

Specifications

Boiling Point 420°C (decomposition)
Melting Point 380°C
Physical Form Powder
Quantity 250 g
UN Number UN2923
Merck Index 14,9125
Solubility Information It reacts with water and ethanol.
Formula Weight 447.27
Percent Purity 99.999%
Packaging Ampule
Sensitivity Moisture sensitive
Density 4.31 g/mL at 15°C
Assay (metals basis)
Chemical Name or Material Tellurium (IV) bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.