missing translation for 'onlineSavingsMsg'
Learn More

Telluric acid dihydrate, 99+%

Product Code. 11367787
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367787 10 g 10g
11357787 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367787 Supplier Thermo Scientific Alfa Aesar Supplier No. 014197.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13451-14-4 | H6O6Te | 229.64 g/mol

Telluric acid dihydrate is used in the preparation of tellurium nanorods through hydrazine-driven reduction, which finds application in solar cells, thermoelectric and topological insulators. It is also involved in the preparation of tellurium metal by reacting with the solution of potassium thiocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13451-14-4
Molecular Formula H6O6Te
Molecular Weight (g/mol) 229.64
MDL Number MFCD00150387
InChI Key VCYUGZHEVVXOPD-UHFFFAOYSA-N
Synonym telluric acid dihydrate, telluricacidwhitepowder, telluric acid-te

Specifications

Boiling Point 160°C (decomposition)
Melting Point 136°C
Color White
Physical Form Crystalline
Quantity 10 g
Assay Percent Range >99%
Solubility Information Soluble in water.
Formula Weight 229.64 (193.61 Anhydrous)
Sensitivity Hygroscopic
Density 3.158
Chemical Name or Material Telluric acid dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.