missing translation for 'onlineSavingsMsg'
Learn More

Kinematica™ Telescopic Electric Stand, ST-P 14/590+500

Product Code. 13497639 Shop All Kinematica Products
Click to view available options
Voltage:
100/120 V
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13497639

Brand: Kinematica™ 11040037

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use this stand with Polytron™ PT 2500 E, PT 4000 and PT 3100 D and PT 6100 D drives. Kinematica™ Telescopic Electric Stand, ST-P 14/590+500 features an ergonomic design and long lasting, robust construction. Includes electric driven stroke, plate and vessel holder.

  • Stand electronics are designed to operate in ambient temperatures of 5 to 40°C and were tested according to EN 60335-1 and UL 962.
  • Ergonomic height adjustment using two buttons
  • Telescopic lift with steel casing
  • Top price-to-performance ratio
  • Also available with vessel holder
  • Powerful integrated lift motor with steel casing
  • Long-lasting, robust construction
  • Excellent stability thanks to solid aluminum base plate
  • Includes non-slip mat
  • Stroke 500mm

Specifications

Product Type Telescopic Electric Stand
Voltage 100/120 V
Length (Metric) 500 mm
Bottom Type Stainless steel base plate
For Use With (Application) Designed to operate in ambient temperatures of 5 to 40°C
For Use With (Equipment) Polytron™ PT 2500 E, PT 4000 and PT 3100 D and PT 6100 D drives.
Model ST-P 14/590+500
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.