missing translation for 'onlineSavingsMsg'
Learn More

Telaglenastat, MedChemExpress

Product Code. 16491277
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491277 50 mg 50mg
16455786 100 mg 100mg
16435048 10 mg 10mg
16487767 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491277 Supplier Medchem Express Supplier No. HY12248
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Telaglenastat (CB-839) is a first-in-class, selective, reversible and orally active glutaminase 1 (GLS1) inhibitor. Telaglenastat selectively inhibits GLS1 splice variants KGA (kidney-type glutaminase) and GAC (glutaminase C) compared to GLS2. The IC50s are 23 nM and 28 nM for endogenous glutaminase in mouse kidney and brain, respectively. Telaglenastat inudces autophagy and has antitumor activity.

Telaglenastat (CB-839) is a first-in-class, selective, reversible and orally active glutaminase 1 (GLS1) inhibitor. Telaglenastat selectively inhibits GLS1 splice variants KGA (kidney-type glutaminase) and GAC (glutaminase C) compared to GLS2. The IC50s are 23 nM and 28 nM for endogenous glutaminase in mouse kidney and brain, respectively. Telaglenastat inudces autophagy and has antitumor activity.

Specifications

Chemical Name or Material Telaglenastat
CAS 1439399-58-2
Color Light Brown
Purity Grade Notes Research
Molecular Formula C26H24F3N7O3S
Quantity 50 mg
Synonym CB-839
Solubility Information DMSO : 6.29 mg/mL (11.00 mM; Need ultrasonic)
SMILES O=C(NC1=NN=C(CCCCC2=NN=C(NC(CC3=CC=CC(OC(F)(F)F)=C3)=O)C=C2)S1)CC4=NC=CC=C4
Molecular Weight (g/mol) 571.57
Formula Weight 571.57
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.