missing translation for 'onlineSavingsMsg'
Learn More

TEL2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18390624 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390624 100 μg 100µL
18399386 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18390624 Supplier Novus Biologicals Supplier No. NBP315520100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TEL2 Polyclonal antibody specifically detects TEL2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEL2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias CLK2, DKFZp434A073, EC 1.1.1.27, EC 2.7.11.1, HCLK2, KIAA0683FLJ10924, Protein clk-2 homolog, TEL2, TEL2, telomere maintenance 2, homolog (S. cerevisiae), telomere length regulation protein TEL2 homolog
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 450-550 of human TEL2 (NP_057195.2).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Direct Reversal of DNA Damage
Primary or Secondary Primary
Gene ID (Entrez) 9894
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.