missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TEKT5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA588509

Product Code. 16576731

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MEFLGTTQTA SYCGPKKCCG LTSLPAVQAP VIQECYQPYY LPGYRYLNSW RPSLFYKIAN VQTCPDESTS TLRPPTILPT LRSALFSRYS PHDWDQSNQL QVRGAEASRL WASRLTDDSM RLLQDKDQLT HQMQEGTCRN LGQRLSDIGF WKSELSYELD RLLTENQNLE TVKRRLECAA NEVNCPLQVA LECLYHREKR IGIDLVHDNV EKNLIREVDL LKCCQEQMRK LAQRIDIQMR DNRDAQHVLE RDLEDKSSAQ CIDEKCFNLR NTSDCISFFH GMEKIDGTIS VPETWAKFSN; Positive Samples: Rat testis; Cellular Location: Cytoplasm, cilium axoneme, cytoskeleton, flagellum axoneme.

May be a structural component of the sperm flagellum. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TEKT5
Polyclonal
Unconjugated
TEKT5
3300001K11Rik; CT149; RGD1307081; Tek5; Tekt5; tektin 5; Tektin5; tektin-5
Rabbit
Affinity Chromatography
RUO
363538, 70426
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.51 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
G5E8A8
TEKT5
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TEKT5 (NP_6532751).
100 μL
Primary
Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.