missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TEKT5 Polyclonal Antibody

Product Code. 12829302
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12829302 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12829302 Supplier Invitrogen™ Supplier No. PA521157

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is NIH-3T3 cell lysate. PA5-21157 can be used with blocking peptide PEP-1271.

May be a structural component of the sperm flagellum. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEKT5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TEKT5
Gene Accession No. G5E8A8, Q96M29
Gene Alias 3300001K11Rik; CT149; RGD1307081; Tek5; Tekt5; tektin 5; Tektin5; tektin-5
Gene Symbols TEKT5
Host Species Rabbit
Immunogen A 17 amino acid peptide near the center of human TEKT5.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146279, 363538, 70426
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.