missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEKT4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531587
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TEKT4 is structural component of ciliary and flagellar microtubules. TEKT4 forms filamentous polymers in the walls of ciliary and flagellar microtubules.
Specifications
| TEKT4 | |
| Polyclonal | |
| Unconjugated | |
| TEKT4 | |
| 1700010L19Rik; RGD1308075; Tek4; Tekt4; tektin 4; Tektin4; tektin-4; Testicular microtubules-related protein 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 150483 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8WW24 | |
| TEKT4 | |
| Recombinant protein encompassing a sequence within the center region of human Tektin 4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction