missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEKT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144737
This item is not returnable.
View return policy
Description
Immunogen sequence: REKRMGIDLV HDEVEAQLLT EVDTILCCQE RMKLHLDKAI AQLAANRASQ HELEKDLSDK QTAYRIDDKC HHLRNTSDGV GYFRGVERVD ATVS Antigen sequence identity: Mouse 83%; Rat 82%.
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. The exact function of this gene is not known. Structural component of ciliary and flagellar microtubules. Forms filamentous polymers in the walls of ciliary and flagellar microtubules. Required for progressive sperm mobility.
Specifications
| TEKT3 | |
| Polyclonal | |
| Unconjugated | |
| TEKT3 | |
| 4933407G07Rik; OMC45; TEKT3; tektin 3; tektin-3; testicular microtubules-related protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 64518 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXF9 | |
| TEKT3 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 202-295. Recombinant protein control fragment (Product #RP-110065). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction