missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEKT2 Monoclonal Antibody (OTI1E6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527237
This item is not returnable.
View return policy
Description
TEKT2 Monoclonal Antibody for Western Blot
Structural component of ciliary and flagellar microtubules. Plays a key role in the assembly or attachment of the inner dynein arm to microtubules in sperm flagella and tracheal cilia. Forms filamentous polymers in the walls of ciliary and flagellar microtubules.
Specifications
| TEKT2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UIF3 | |
| TEKT2 | |
| Human recombinant protein fragment corresponding to amino acids 1-244 of TEKT2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1E6 | |
| Unconjugated | |
| TEKT2 | |
| AU014974; h-tektin-t; TEKT2; TEKTB1; tektin 2; tektin 2 (testicular); tektin-2; tektin-B1; TEKTIN-T; Testicular tektin; testicular tektin B1-like protein | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27285 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction