Learn More
Teknon™ Biochlor Effervescent Chlorine Tablets
Suitable for surface cleaning and reagent preparation. Teknon™ Biochlor Effervescent Chlorine Tablets primary application is solubilizing hydrophobic molecules.
Brand: Teknon TK290
3.22 GBP valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Code : 24
Additional Details : CAS Number : 124-04-9 Weight : 0.20000kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Description
- Suitable for general cleaning, disinfecting and washing
- Fast dissolving
- Kills bacteria, fungi, viruses and spores
- Effective, economical and convenient
Specifications
Chlorine Tablets | |
White | |
200 Tablets |
Safety and Handling
- Biochlor
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P391-Collect spillage.
- P402+P404-Store in a dry place. Store in a closed container.
- P405-Store locked up.
- P501-Dispose of contents/container to .