missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TEK (TIE2) [R849W], GST Tag, His Tag Recombinant Protein

Product Code. 15401214
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15401214

Brand: Invitrogen™ PV6226

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Human TEK (TIE2) R849W, C-terminal fragment, amino acids Q771-A1124 (as in NCBI/Protein entry NP_000450.1) with a R849W mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Related Product: LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant

TIE2, also known as TEK tyrosine kinase, TIE2 gene is mapped to 9p21.2. This gene encodes a receptor that belongs to the protein tyrosine kinase Tie2 family. The encoded protein possesses a unique extracellular region that contains two immunoglobulin-like domains, three epidermal growth factor (EGF)-like domains and three fibronectin type III repeats. The ligand angiopoietin-1 binds to this receptor and mediates a signaling pathway that functions in embryonic vascular development. Immunoblotting showed that TIE2 expression was increased by thyroid-stimulating hormone and agents that increased intracellular cAMP. HSCs expressing the receptor tyrosine kinase TIE2 are quiescent and antiapoptotic and comprise a side population of HSCs that adhere to osteoblasts in the bone marrow niche.
TRUSTED_SUSTAINABILITY

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM HEPES with 100 mM NaCl, 15 mM glutathione, 20% glycerol, 5 mM DTT and no preservative; pH 7.5
Molecular Weight (g/mol) 69 kDa
Name Human TEK (TIE2) [R849W], GST Tag, His Tag
Quantity 10 μg
Common Name TIE2 (TEK) (R849W Mutant)
Protein Length 771-1124
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag, His-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.