missing translation for 'onlineSavingsMsg'
Learn More

Teicoplanin, Thermo Scientific™

Product Code. 15565896 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15565896 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15565896 Supplier Thermo Scientific Acros Supplier No. 455400010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Type Teicoplanin
Physical Form Powder
CAS 61036-62-2
Melting Point 260.0°C
Color White to Yellow
Quantity 1 g
Molecular Formula C88H97Cl2N9O33
MDL Number MFCD05664625
Synonym Targocid, Teichomycin
InChI Key BJNLLBUOHPVGFT-QRZIFLFXSA-N
SMILES CCCCCCCCCC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OC1=C2OC3=CC=C(C[C@H]4NC(=O)[C@H](N)C5=CC=C(O)C(OC6=CC(O)=CC(=C6)[C@H](NC4=O)C(=O)N[C@@H]4C(C=C1OC1=CC=C(C=C1Cl)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(C)=O)[C@@H]1NC(=O)[C@H](NC4=O)C4=CC=C(O)C(=C4)C4=C(O[C@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]6O)C=C(O)C=C4[C@@H](NC1=O)C(O)=O)=C2)=C5)C=C3Cl
IUPAC Name (1S,2R,19R,22R,34S,37R,40R,52R)-22-amino-63,65-dichloro-64-{[(2S,3R,4R,5S,6R)-3-decanamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-{[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-26,31,44,49-tetrahydroxy-21,35,38,54,56,59-hexaoxo-47-{[(2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-7,13,28-trioxa-20,36,39,53,55,58-hexaazaundecacyclo[38.14.2.2³,⁶.2¹⁴,¹⁷.2¹⁹,³⁴.1⁸,¹².1²³,²⁷.1²⁹,³³.1⁴¹,⁴⁵.0¹⁰,³⁷.0⁴⁶,⁵¹]hexahexaconta-3,5,8,10,12(64),14,16,23,25,27(61),29,31,33(60),41,43,45(57),46,48,50,62,65-henicosaene-52-carboxylic acid
Molecular Weight (g/mol) 1879.67
PubChem CID 133640186
Chemical Name or Material Teicoplanin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.