missing translation for 'onlineSavingsMsg'
Learn More

Sonoco™ Tegrant ThermoSafe LabMailer Absorbent Sheets

Product Code. p-4919710
Click to view available options
Capacity:
22.5 mL of blood/sheet
Diameter (Metric) Interior:
7.6 x 7.6 cm
Diameter (English) Interior:
3 x 3 in.
Unit Size:
Pack of 250
This item is not returnable. View return policy

Product Code. 13501930

Brand: Sonoco™ Zorb33

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lightweight, economical lab mailer shippers

  • Accommodates VacuTainer™, Microtainer™, and other collection/transport tubes
  • Available in multi-tube, slide, and tube/slide combinations
  • Polystyrene foam construction saves postage
  • Reusable and recyclable

Additional packaging may be required to conform to CFR72.2 specification (See Packaging Accessories Kit Mfr. No. 471).

TRUSTED_SUSTAINABILITY

Specifications

Quantity 22.5mL of blood per sheet
For Use With (Equipment) Provide maximum breakage protection for known non-hazardous materials.
Applications For protection of frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use.
Diameter (English) Interior 3 x 3 in.
Diameter (Metric) Interior 7.6 x 7.6 cm
Capacity 22.5 mL of blood/sheet
Material Lightweight Polystyrene Foam
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.