missing translation for 'onlineSavingsMsg'
Learn More

Tegafur, MedChemExpress

Product Code. 16481846
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481846 500 mg 500mg
16427286 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481846 Supplier Medchem Express Supplier No. HY17400
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tegafur (FT 207; NSC 148958) is a chemotherapeutic 5-FU prodrug used in the treatment of cancers; is a component of tegafur-uracil.

Tegafur (FT 207

Specifications

Chemical Name or Material Tegafur
CAS 17902-23-7
Color White
Purity Grade Notes Research
Molecular Formula C8H9FN2O3
Quantity 500 mg
Synonym FT 207 NSC 148958
Solubility Information DMSO : ≥ 48 mg/mL (239.80 mM)
SMILES O=C1NC(C(F)=CN1C2OCCC2)=O
Molecular Weight (g/mol) 200.17
Formula Weight 200.17
Percent Purity 98.74%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.