Learn More
Invitrogen™ TEFM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597096
Description
Immunogen sequence: KSTTPKKITP NVTFCDENAK EPENALDKLF SSEQQASILH VLNTASTKEL EAFRLLRGRR SINIVEHREN FGPFQNLESL MNVPLFKYKS TVQVCNSILC PKTGREKRKS PENRFLRKLL KPDIERERLK AVNSIISIVF GTRRIAWAHL DRKLTVLDWQ QSDRWSLMRG IYSSS; Positive Samples: HL-60, K-562, Mouse liver, Rat liver; Cellular Location: Mitochondrion matrix, mitochondrion nucleoid.
TEFM is a transcription elongation factor which increases mitochondrial RNA polymerase processivity. It regulates transcription of the mitochondrial genome, including genes important for the oxidative phosphorylation machinery.
Specifications
| TEFM | |
| Polyclonal | |
| Unconjugated | |
| TEFM | |
| 1110002N22Rik; C17orf42; C9H17orf42; RGD1306595; Tefm; transcription elongation factor of mitochondria; transcription elongation factor, mitochondrial | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 287554, 68550, 79736 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.9 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4KM51, Q5SSK3, Q96QE5 | |
| TEFM | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.