missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEFM Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£248.00 - £423.00
Specifications
| Antigen | TEFM |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18481511
|
Novus Biologicals
NBP1-82109-25ul |
25 μL |
£248.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18250586
|
Novus Biologicals
NBP1-82109 |
0.1 mL | |||||||
Description
TEFM Polyclonal specifically detects TEFM in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TEFM | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C17orf42, chromosome 17 open reading frame 42, FLJ22729, MGC24674, TEFM, transcription elongation factor of mitochondria, transcription elongation factor, mitochondrial | |
| TEFM | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 79736 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RRSINIVEHRENFGPFQNLESLMNVPLFKYKSTVQVCNSILCPKTGREKRKSPENRFLRKLLKPDIERERLKAVNSIISIV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title