missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEF5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539575
This item is not returnable.
View return policy
Description
This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is predomitly expressed in the placenta and is involved in the transactivation of the chorionic somatomammotropin-B gene enhancer. Translation of this protein is initiated at a non-AUG (AUA) start codon. [provided by RefSeq, Jul 2008].
Specifications
| TEF5 | |
| Polyclonal | |
| Unconjugated | |
| TEAD3 | |
| TEAD5, TEF5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21678, 7005 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99594, P70210 | |
| TEAD3 | |
| A synthetic peptide derived from the internal region of human TEAD3 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction