missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556008
This item is not returnable.
View return policy
Description
Immunogen sequence: QDIKPFAQPA YPIQPPLPPT LSSYEPLAPL PSAAASVPVW QDRTIASSRL RLLEYSAFME VQRDPDTYSK HLFVHIGQTN PAFSDPPLEA VDVRQIYDKF PEK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.
This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is predominantly expressed in the placenta and is involved in the transactivation of the chorionic somatomammotropin-B gene enhancer. Translation of this protein is initiated at a non-AUG (AUA) start codon.
Specifications
| TEF5 | |
| Polyclonal | |
| Unconjugated | |
| TEAD3 | |
| DTEF-1; ETFR-1; ETF-related factor 1; Tcf13r2; TEA domain family member 3; TEA domain family member 5; TEA domain transcription factor 3; Tead3; TEAD-3; TEAD5; TEF5; TEF-5; transcriptional enhancer factor 5; transcriptional enhancer factor TEF-5; transcriptional enhancer factor TEF-5 (DTEF-1) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7005 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99594 | |
| TEAD3 | |
| Recombinant protein corresponding to Human TEF5. Recombinant protein control fragment (Product #RP-102354). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction