missing translation for 'onlineSavingsMsg'
Learn More

TEF1 Rabbit anti-Human, Mouse, Rat, Clone: 2E7N10, Novus Biologicals™

Product Code. 18361135 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361135 20 μg 20µL
18383906 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18361135 Supplier Novus Biologicals Supplier No. NBP31659320UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TEF1 Monoclonal antibody specifically detects TEF1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEF1
Applications ChIP Assay, Immunoprecipitation
Classification Monoclonal
Clone 2E7N10
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias NTEF-1, protein GT-IIC, TEA domain family member 1 (SV40 transcriptional enhancer factor), TEAD-1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TEF1 (P28347).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 7003
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.