missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP BIOMEDICALS TeenPrep Adapter Bottom Component
Shop All MP Biomedicals Products
Click to view available options
For Use With (Equipment):
FastPrep-24
Unit Size:
Each
Description
- Bottom of TeenPrep Adapter for FastPrep-24 instrument
- Holds 12 15-mL tubes
- For room temperature lysis and grinding of samples
Specifications
Specifications
| Product Type | Teen Prep Adapter |
| For Use With (Equipment) | FastPrep-24 |
| Holds | 12 Tubes (15 mL) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction