Learn More
VICI Valco Instruments™ Tee 10um titanium frit
Product Code. 12915001
Product Code. 12915001
Brand: VICI Valco Instruments™ JR90000670
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
350bar (5000psi) PEEK internal volume including frit: 4μL body dimensions: 25.4mm x 9.5mm thick tolerances: +- 0.05mm (0.002inch)