missing translation for 'onlineSavingsMsg'
Learn More

Teduglutide Trifluoroacetic Acid Salt, TRC

Product Code. 30011615
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011615 10 mg 10mg
30004231 1 mg 1mg
30005672 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011615 Supplier Toronto Research Chemicals Supplier No. TRCT13800010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C164H252N44O55S • x(C2HF3O2)
Synonym 1: PN: US20130072430 SEQID: 4 Claimed Protein TFA Salt, ALX 0600 TFA Salt, Gattex TFA Salt, Glucagon-like Peptide II [2-Glycine] (Human) TFA Salt, Revestive TFA Salt;
SMILES [H]N[C@@H](CC1=CN=CN1)C(NCC(N[C@@H](CC(O)=O)C(NCC(N[C@H](C(N[C@@H](CC2=CC=CC=C2)C(N[C@H](C(N[C@@H](CC(O)=O)C(N[C@H](C(N[C@@H](CCSC)C(N[C@H](C(N[C@H](C(N[C@@H]([C@H](CC)C)C(N[C@H](C(N[C@@H](CC(O)=O)C(N[C@H](C(N[C@H](C(N[C@@H](C)C(N[C@@H](C)C(N[C@H](C(N[C@@H](CC(O)=O)C(N[C@@H](CC3=CC=CC=C3)C(N[C@@H]([C@H](CC)C)C(N[C@H](C(N[C@@H](CC4=CNC5=C4C=CC=C5)C(N[C@H](C(N[C@@H]([C@H](CC)C)C(N[C@H](C(N[C@H](C(N[C@H](C(N[C@@H]([C@H](CC)C)C(N[C@H](C(N[C@@H](CC(O)=O)C(O)=O)=O)[C@@H](C)O)=O)=O)CCCCN)=O)[C@@H](C)O)=O)CCC(N)=O)=O)=O)CC(C)C)=O)=O)CC(N)=O)=O)=O)=O)=O)CCCNC(N)=N)=O)=O)=O)CC(C)C)=O)CC(N)=O)=O)=O)CC(C)C)=O)=O)[C@@H](C)O)=O)CC(N)=O)=O)=O)CCC(O)=O)=O)=O)CO)=O)=O)CO)=O)=O)=O)=O.O=C(C(F)(F)F)O

Specifications

Shipping Condition Room Temperature
InChI Formula InChI=1S/C164H252N44O55S.C2HF3O2/c1-21-77(11)126(156(255)187-95(44-46-114(167)214)141(240)206-130(83(17)211)160(259)186-93(42-33-34-49-165)140(239)202-129(80(14)24-4)159(258)208-131(84(18)212)161(260)200-111(163(262)263)66-125(230)231)203-151(250)100(54-76(9)10)189-145(244)103(57-88-67-175-92-41-32-31-40-90(88)92)192-147(246)105(60-116(169)216)199-157(256)127(78(12)22-2)204-152(251)102(56-87-38-29-26-30-39-87)190-149(248)109(64-123(226)227)195-137(236)94(43-35-50-174-164(171)172)183-134(233)82(16)179-133(232)81(15)180-142(241)98(52-74(5)6)188-146(245)104(59-115(168)215)194-150(249)110(65-124(228)229)196-143(242)99(53-75(7)8)198-158(257)128(79(13)23-3)205-162(261)132(85(19)213)207-153(252)106(61-117(170)217)193-139(238)97(48-51-264-20)185-138(237)96(45-47-120(220)221)184-148(247)108(63-122(224)225)197-155(254)113(72-210)201-144(243)101(55-86-36-27-25-28-37-86)191-154(253)112(71-209)182-119(219)70-177-136(235)107(62-121(222)223)181-118(218)69-176-135(234)91(166)58-89-68-173-73-178-89;3-2(4,5)1(6)7/h25-32,36-41,67-68,73-85,91,93-113,126-132,175,209-213H,21-24,33-35,42-66,69-72,165-166H2,1-20H3,(H2,167,214)(H2,168,215)(H2,169,216)(H2,170,217)(H,173,178)(H,176,234)(H,177,235)(H,179,232)(H,180,241)(H,181,218)(H,182,219)(H,183,233)(H,184,247)(H,185,237)(H,186,259)(H,187,255)(H,188,245)(H,189,244)(H,190,248)(H,191,253)(H,192,246)(H,193,238)(H,194,249)(H,195,236)(H,196,242)(H,197,254)(H,198,257)(H,199,256)(H,200,260)(H,201,243)(H,202,239)(H,203,250)(H,204,251)(H,205,261)(H,206,240)(H,207,252)(H,208,258)(H,220,221)(H,222,223)(H,224,225)(H,226,227)(H,228,229)(H,230,231)(H,262,263)(H4,171,172,174);(H,6,7)/t77-,78-,79-,80-,81-,82-,83+,84+,85+,91-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,113-,126-,127-,128-,129-,130-,131-,132-;/m0./s1
Industry Type Pharmaceutical
Quantity 10 mg
Chemical Name or Material Teduglutide Trifluoroacetic Acid Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.