missing translation for 'onlineSavingsMsg'
Learn More

Tedatioxetine hydrobromide, MedChemExpress

Product Code. 16436277
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16436277 5 mg 5mg
16403558 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16436277 Supplier Medchem Express Supplier No. HY101755
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tedatioxetine (Lu AA24530) hydrobromide acts as a serotonin and norepinephrine (NE)-preferring triple reuptake inhibitor (TRI) and 5-HT2A, 5-HT2C, 5-HT3 and α1A-adrenergic receptor antagonist. ,

Tedatioxetine (Lu AA24530) hydrobromide acts as a serotonin and norepinephrine (NE)-preferring triple reuptake inhibitor (TRI) and 5-HT2A, 5-HT2C, 5-HT3 and α1A-adrenergic receptor antagonist.

Specifications

Chemical Name or Material Tedatioxetine hydrobromide
CAS 960151-65-9
Color White
Purity Grade Notes Research
Molecular Formula C18H22BrNS
Quantity 5 mg
Synonym Lu AA24530 hydrobromide
Solubility Information DMSO : 100 mg/mL (274.47 mM; Need ultrasonic)
SMILES CC1=CC=C(SC2=CC=CC=C2C3CCNCC3)C=C1.[H]Br
Molecular Weight (g/mol) 364.34
Formula Weight 364.34
Percent Purity 98.44%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.