missing translation for 'onlineSavingsMsg'
Learn More

Tectochrysin, MedChemExpress

Product Code. 16438707
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438707 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438707 Supplier Medchem Express Supplier No. HY14592
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tectochrysin (Techtochrysin) is one of the major flavonoids of Alpinia oxyphylla Miquel. Tectochrysin inhibits activity of NF-κB.

Tectochrysin (Techtochrysin) is one of the major flavonoids of Alpinia oxyphylla Miquel. Tectochrysin inhibits activity of NF-κB.

Specifications

Chemical Name or Material Tectochrysin
CAS 520-28-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H12O4
Quantity 5 mg
Synonym Techtochrysin NSC 80687
Solubility Information DMSO : 10 mg/mL (37.28 mM; Need ultrasonic)
SMILES OC1=C2C(OC(C3=CC=CC=C3)=CC2=O)=CC(OC)=C1
Molecular Weight (g/mol) 268.26
Formula Weight 268.26
Percent Purity 99.16%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.