missing translation for 'onlineSavingsMsg'
Learn More

Tecnazene 100 μg/mL in Ethanol, Dr. Ehrenstorfer

Product Code. 30324416
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324416 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324416 Supplier Dr. Ehrenstorfer Supplier No. DREA17200000EL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3,5,6-Tetrachloronitrobenzene
CAS 117-18-0
Analyte or Component Names 2,3,5,6-Tetrachloronitrobenzene
Shipping Condition +20°C
Molecular Formula C6 H Cl4 N O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2,3,5,6-Tetrachloronitrobenzene, 1,2,4,5-Tetrachloro-3-nitrobenzene, 1-Nitro-2,3,5,6-tetrachlorobenzene, 2,3,5,6-Tetrachloro-1-nitrobenzene, Chipman 3142, Folosan, Folosan DB 905, Fusarex, Myfusan, NSC 10235, Napotate, Nebulin, TCNB, TCNB (pesticide), Tecnazen, Tecnazene, Tetrachloronitrobenzene
InChI Formula InChI=1S/C6HCl4NO2/c7-2-1-3(8)5(10)6(4(2)9)11(12)13/h1H
SMILES [O-][N+](=O)c1c(Cl)c(Cl)cc(Cl)c1Cl
IUPAC Name 1,2,4,5-tetrachloro-3-nitrobenzene
Solvent or Matrix Ethanol
Molecular Weight (g/mol) 260.89
Quantity 1 mL
Formula Weight 258.8761 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.