missing translation for 'onlineSavingsMsg'
Learn More

Tecloftalam Imide, TRC

Product Code. 30426816
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426816 10 mg -
30426815 25 mg 25mg
30411424 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426816 Supplier Toronto Research Chemicals Supplier No. TRCT01371010MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26491-30-5
Molecular Formula C14 H3 Cl6 N O2
Molecular Weight (g/mol) 429.9
Synonym 4,5,6,7-Tetrachloro-2-(2,3-dichlorophenyl)-1H-isoindole-1,3(2H)-dione (ACI), Phthalimide, 3,4,5,6-tetrachloro-N-(2,3-dichlorophenyl)- (8CI), Tecloftalam-imide
IUPAC Name 4,5,6,7-tetrachloro-2-(2,3-dichlorophenyl)isoindole-1,3-dione
SMILES Clc1cccc(N2C(=O)c3c(Cl)c(Cl)c(Cl)c(Cl)c3C2=O)c1Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H3Cl6NO2/c15-4-2-1-3-5(8(4)16)21-13(22)6-7(14(21)23)10(18)12(20)11(19)9(6)17/h1-3H
Formula Weight 426.8295
Chemical Name or Material Tecloftalam Imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.