Learn More
Invitrogen™ TECK Monoclonal Antibody (89818)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA523847
Description
In direct ELISAs, no cross-reactivity with recombinant mouse (rm) CCL1, 2, 3, 4, 5, 6, 7, 9, 11, 12, 17, 19, 20, 21, 22 or 24, recombinant human (rh) CCL2, 3, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25 or 26, or recombinant rat (rr) CCL20 is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Chemokines are likely to play an important role in regulating the trafficking of developing T cells within the thymus. Chemokine C-C thymus expressed chemokine (TECK), also designated chemokine ligand 25 (CCL25), small induc-ible cytokine A25, chemokine b-15 or CK b-15, is expressed predominantly in thymic dendritic cells, thymic epithelial cells and in the small intestine. TECK, a CCR9 ligand, has suppressive activity against immature subsets of myeloid progenitors which have been stimulated to proliferate by multiple growth factors. TECK delivers signals through CCR9, which is important for the nav-igation of developing thymocytes. Bone marrow pre-pro-B cells and cells capable of generating pro-B colonies in the presence of interleukin-7 and Flt-3 ligand migrate to TECK, a response lost in later stages of B cell development.
Specifications
| TECK | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O35903 | |
| CCL25 | |
| E. coli-derived recombinant mouse CCL25/TECK Gln24-Asn144. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Neutralization, Western Blot | |
| 89818 | |
| Unconjugated | |
| CCL25 | |
| A130072A22Rik; AI852536; C-C motif chemokine 25; C-C motif chemokine ligand 25; CCL25; chemokine (C-C motif) ligand 25; chemokine TECK; Ck beta-15; CKb15; Scya25; SCYA25TECKvar; small inducible chemokine 25; small inducible cytokine A25; small inducible cytokine subfamily A (Cys-Cys), member 25; small-inducible cytokine A25; Teck; TECKMGC150327; TECKvar; thymus expressed chemokine; thymus-expressed chemokine | |
| Rat | |
| Protein A/G | |
| RUO | |
| 20300 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.