Learn More
Invitrogen™ Mouse TECK (CCL25) ELISA Kit
Description
The Mouse TECK (CCL25) ELISA quantitates Ms TECK in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms TECK. Principle of the method The Mouse TECK solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | A130072A22Rik; AI852536; C-C motif chemokine 25; C-C motif chemokine ligand 25; Ccl25; chemokine (C-C motif) ligand 25; chemokine TECK; Ck beta-15; CKb15; Scya25; SCYA25TECKvar; small inducible chemokine 25; small inducible cytokine A25; small inducible cytokine subfamily A (Cys-Cys), member 25; Small-inducible cytokine A25; Teck; TECKMGC150327; TECKvar; thymus expressed chemokine; thymus-expressed chemokine |
| Assay Range | 20.5 to 2000 pg/mL |
| Assay Sensitivity | 15 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 20300 |
| Gene Symbol | CCL25 |
| Interassay CV | <12% |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.