Learn More
BRAND™ Technical Quality PFA Bottle
Product Code. 10530003 Shop All Brand Products
Click to view available options
Capacity (Metric):
100 mL
500 mL
Unit Size:
Each
Product Code. 10530003
Brand: BRAND™ 130486
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Technical Quality PFA Bottle includes an ETFE screw cap and narrow mouth.
  • With screw caps made of ETFE
  • These bottles contain a small fraction of reprocessed PFA from our production process
  • The physical properties and chemical resistance compare with high purity PFA
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Material PFA
Mouth Narrow
Closure Material ETFE
Closure Type Screw Cap