missing translation for 'onlineSavingsMsg'
Learn More

Technical Calibration Buffer pH 1.68, Hanna Instruments™

Product Code. 11755055
Click to view available options
Quantity:
20 mL
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 11755055

Brand: Hanna Instruments HI5001601

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Colorless
pH 1.68
Physical Form Liquid
Quantity 20 mL
Solution Type Buffer
Formulation Aqueous Buffer Solution
Odor Odorless
compliance-icons
Product Identifier
  • pH 1.68 Buffer Solution
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Potassium tetraoxalate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.