missing translation for 'onlineSavingsMsg'
Learn More

Technical Buffer Sachets, Mettler Toledo™
GREENER_CHOICE

Product Code. 15339400 Shop All Mettler Toledo Products
Change view
Click to view available options
Quantity:
30 x 20 mL
Unit Size:
Pack of 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15339400 30 x 20 mL Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15339400 Supplier Mettler Toledo Supplier No. 30111134

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Traceable to the primary standards of reference materials selected by NIST.

  • Sachets guarantee fresh solution for every calibration
  • Easy to use
TRUSTED_SUSTAINABILITY

Specifications

pH 2
Quantity 30 x 20 mL
Solution Type Buffer
Packaging Sachets
compliance-icons
Product Identifier
  • buffer solution pH 2.00
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: citric acid anhydride, hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.