missing translation for 'onlineSavingsMsg'
Learn More

Technical Buffer Solution pH 7, WTW™

Product Code. 10216304
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Packaging:
1 Bottle
10 Bottles
25 Bottles
Unit Size:
250mL
Each
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10216304 250 mL 25 Bottles Pack of 25
11745164 250 mL 1 Bottle 250mL
10595505 1000 mL 10 Bottles Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10216304 Supplier WTW Supplier No. 108812
  • ONLINE EXCLUSIVE PRICE
    £347.00
    £339.41 / Pack of 25
    Save £7.59
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Precise and traceable to NIST (National Institute of Standards and Technology), their accuracy is ± 0,03 pH.

  • Precise and traceable to NIST (National Institute of Standards and Technology, USA in bulk size of 250 ml with built-in dosing vessel standard buffer
  • Life up to 3 years (unopened)
  • Thrifty use
  • Easiest handling by means of dosing device
TRUSTED_SUSTAINABILITY

Specifications

pH 7
Quantity 250 mL
Packaging 25 Bottles
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.